Learn More
Selleck Chemical LLCSupplier Diversity Partner Bicalutamide 100mg 90357-06-5 ICI-176334  DFS Item
Manufacturer:  Selleck Chemical LLCSupplier Diversity Partner S1190100MG
Bicalutamide (ICI-176334) is an androgen receptor (AR) antagonist with IC50 of 0.16 μM in LNCaP/AR(cs)cell line. Bicalutamide promotes autophagy.
Catalog No. 50-624-0