Learn More
Selleck Chemical LLC
Bicalutamide 200mg 90357-06-5 ICI-176334
Manufacturer: Selleck Chemical LLC S1190200MG
Bicalutamide (ICI-176334) is an androgen receptor (AR) antagonist with IC50 of 0.16 μM in LNCaP/AR(cs)cell line. Bicalutamide promotes autophagy.