Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BICD2 Mouse anti-Human, Mouse, Rat, HRP, Clone: 10811, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP243683H

Catalog No. NBP243683H


Add to cart

Description

Description

BICD2 Monoclonal antibody specifically detects BICD2 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications

Specifications

BICD2
10811
Western Blot
Purified
BICD2
Recombinant protein encompassing a sequence within the N-terminus region of human BICD2. The exact sequence is proprietary.
Protein A purified
Store at 4C in the dark.
Monoclonal
Human, Mouse, Rat
Western Blot
HRP
PBS with No Preservative
bicaudal D homolog 2 (Drosophila), KIAA0699
Mouse
IgG2b
0.1 ml
Primary
23299
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit