Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Bicine, 0.2M buffer soln., pH 9.0, low endotoxin

Click to view available options
Quantity:
100mL
250mL
150-25-4
C6H13NO4
163.173
MFCD00004295
FSVCELGFZIQNCK-UHFFFAOYSA-N
8761
CHEBI:40957
2-[bis(2-hydroxyethyl)amino]acetic acid
C(CO)N(CCO)CC(=O)O
150-25-4
C6H13NO4
163.173
MFCD00004295
FSVCELGFZIQNCK-UHFFFAOYSA-N
8761
CHEBI:40957
2-[bis(2-hydroxyethyl)amino]acetic acid
C(CO)N(CCO)CC(=O)O
Concentration 0.2M buffer soln.
Physical Form Liquid
pH 9.0
Color Colorless
Quantity 100mL
Chemical Name or Material Bicine, low endotoxin

TSCA : TSCA

storageNote1 : 2° to 8°C