Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Bicine, 98.5 to 101.5% (Dry Basis), Ultrapure, Thermo Scientific™

Click to view available options
Quantity:
100g
1kg
Packaging:
Plastic bottle
150-25-4
C6H13NO4
163.173
MFCD00004295
FSVCELGFZIQNCK-UHFFFAOYSA-N
N,N-bis[2-Hydroxyethyl]glycine
8761
CHEBI:40957
2-[bis(2-hydroxyethyl)amino]acetic acid
C(CO)N(CCO)CC(=O)O
150-25-4
C6H13NO4
163.173
MFCD00004295
FSVCELGFZIQNCK-UHFFFAOYSA-N
N,N-bis[2-Hydroxyethyl]glycine
8761
CHEBI:40957
2-[bis(2-hydroxyethyl)amino]acetic acid
C(CO)N(CCO)CC(=O)O
Linear Formula (CH2CH2OH)2NCH2COOH
Formula Weight 163.17
Physical Form Crystalline Powder
Percent Purity 98.5 to 101.5%
Packaging Plastic bottle
Melting Point ~191°C (decomposition)
Quantity 100g
Chemical Name or Material Bicine
Assay Dry basis