Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bicine, 99+%, for analysis, ACROS Organics™

Click to view available options
Quantity:
100g
250g
1kg
Packaging:
Glass bottle
150-25-4
C6H13NO4
163.173
MFCD00004295
FSVCELGFZIQNCK-UHFFFAOYSA-N
N, N-Bis(2-hydroxyethyl)glycine,Diethylolglycine
8761
CHEBI:40957
2-[bis(2-hydroxyethyl)amino]acetic acid
C(CO)N(CCO)CC(=O)O
150-25-4
C6H13NO4
163.173
MFCD00004295
FSVCELGFZIQNCK-UHFFFAOYSA-N
N, N-Bis(2-hydroxyethyl)glycine,Diethylolglycine
8761
CHEBI:40957
2-[bis(2-hydroxyethyl)amino]acetic acid
C(CO)N(CCO)CC(=O)O
Biological buffer
Absorbance 0.05 max. (5% aqueous soln.) at 250nm
Absorbance Conditions (5% aq. soln.) (1cm cell)
CAS Min % 99.0
CAS Max % 100.0
Chloride 50ppm max.
Chlorine (Cl) 50ppm max.
Assay Percent Range 99+%
Linear Formula (HOCH2CH2)2NCH2CO2H
Copper (Cu) 2ppm max.
Merck Index 15, 1205
Heavy Metals (as Pb) 5ppm max.
Formula Weight 163.17
Physical Form Crystalline Powder
Percent Purity ≥99.0% (Total base)
Grade Analysis
Infrared Spectrum Authentic
Lead (Pb) 5ppm max.
Iron (Fe) 5ppm max.
Packaging Glass bottle
Sulfate 50ppm max.
Sulfated Ash 0.1 % max.
Water 1% Max. (K.F.)
pH 4.0 to 5.5 (10% soln. at 20°C)
Color White
Quantity 100g
Chemical Name or Material Bicine, 99+%
Show More Show Less

EINECSNumber : 205-755-1

RTECSNumber : MB9700000

TSCA : TSCA