Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ (-)-Bicuculline methobromide

Click to view available options
Quantity:
10 mg
50 mg
Description
Methobromide salt of (+)-bicuculline . Water soluble and more stable than bicuculline. Non-GABA receptor-mediated actions reported.
Methoiodide Salt and Methochloride Salt also available.
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 462.3 |
Purity | >98% |
Molecular Weight (g/mol) | 462.296 |
Chemical Name or Material | (-)-Bicuculline methobromide |
Safety and Handling
Recommended Storage | Store at Room Temperature |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction