Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(-)-Bicuculline methobromide, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
10mg
50mg

Chemical Identifiers

73604-30-5
C21H20BrNO6
462.296
BWXCECYGGMGBHD-UHFFFAOYSA-M
[R-(R™,S™)]-5-(6,8-Dihydro-8-oxofuro[3,4-e]-1,3-benzodioxol-6-yl)-5,6,7,8-tetrahydro-6,6-dimethyl-1,3-dioxolo[4,5-g]isoquinolinium bromide
171729
6-(6,6-dimethyl-7,8-dihydro-5H-[1,3]dioxolo[4,5-g]isoquinolin-6-ium-5-yl)-6H-furo[3,4-g][1,3]benzodioxol-8-one;bromide
C[N+]1(CCC2=CC3=C(C=C2C1C4C5=C(C6=C(C=C5)OCO6)C(=O)O4)OCO3)C.[Br-]

Water-soluble GABAA antagonist

Methobromide salt of (+)-bicuculline (Cat. No. 0130). Water soluble and more stable than bicuculline. Non-GABA receptor-mediated actions reported.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Quantity 10mg
Formula Weight 462.3
Chemical Name or Material (-)-Bicuculline methobromide
Grade >98%

Recommended Storage : Store at Room Temperature