Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bicyclo[2.2.2]oct-7-ene-2,3,5,6-tetracarboxylic dianhydride, 97%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
1719-83-1
C12H8O6
248.19
MFCD00082228
XLOGCGOPKPCECW-UHFFFAOYSA-N
bicyclo 2.2.2 oct-7-ene-2,3,5,6-tetracarboxylic dianhydride,4,4a,8,8a-tetrahydro-4,8-ethenobenzo 1,2-c:4,5-c' difuran-1,3,5,7 3ah,7ah-tetraone,bicyclo 2.2.2 oct-7-ene-2,3,5,6-tetracarboxylic acid dianhydride,4,8-etheno-1h,3h-benzo 1,2-c:4,5-c' difuran-1,3,5,7-tetrone, 3a,4,4a,7a,8,8a-hexahydro,bicyclo 2.2.2-7-octene-2,3,5,6-tetracarboxylic dianhydride,bicyclo 2.2.2-7-octene-2,3,5,6-tetracarboxylic acid dianhydride,4,10-dioxatetracyclo 5.5.2.0<2,6>.0<8,12> tetradec-13-ene-3,5,9,11-tetraone,4,10-dioxatetracyclo 5.5.2.0 2 ,?.0?, 1 2 tetradec-13-ene-3,5,9,11-tetrone,bicyclo 2.2.2 oct-7-ene-2,3:5,6-tetracarboxylic dianhydride,4,4a,8,8a-tetrahydro-4,8-ethenobenzo 1,2-c
102679
C1=CC2C3C(C1C4C2C(=O)OC4=O)C(=O)OC3=O
1719-83-1
C12H8O6
248.19
MFCD00082228
XLOGCGOPKPCECW-UHFFFAOYSA-N
bicyclo 2.2.2 oct-7-ene-2,3,5,6-tetracarboxylic dianhydride,4,4a,8,8a-tetrahydro-4,8-ethenobenzo 1,2-c:4,5-c' difuran-1,3,5,7 3ah,7ah-tetraone,bicyclo 2.2.2 oct-7-ene-2,3,5,6-tetracarboxylic acid dianhydride,4,8-etheno-1h,3h-benzo 1,2-c:4,5-c' difuran-1,3,5,7-tetrone, 3a,4,4a,7a,8,8a-hexahydro,bicyclo 2.2.2-7-octene-2,3,5,6-tetracarboxylic dianhydride,bicyclo 2.2.2-7-octene-2,3,5,6-tetracarboxylic acid dianhydride,4,10-dioxatetracyclo 5.5.2.0<2,6>.0<8,12> tetradec-13-ene-3,5,9,11-tetraone,4,10-dioxatetracyclo 5.5.2.0 2 ,?.0?, 1 2 tetradec-13-ene-3,5,9,11-tetrone,bicyclo 2.2.2 oct-7-ene-2,3:5,6-tetracarboxylic dianhydride,4,4a,8,8a-tetrahydro-4,8-ethenobenzo 1,2-c
102679
C1=CC2C3C(C1C4C2C(=O)OC4=O)C(=O)OC3=O
Elemental Analysis 56.00 to 60.10% (C)
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 100g
Color White to Beige
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Formula Weight 248.19
Physical Form Crystals or Crystalline Powder
Chemical Name or Material Bicyclo[2.2.2]oct-7-ene-2,3,5,6-tetracarboxylic dianhydride, 98%
Show More Show Less

GHS P Statement
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

MOISTURE SENSITIVE

EINECSNumber : 217-009-2