Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bicyclo[3.2.0]hept-2-en-6-one, 97%, Alfa Aesar™

Click to view available options
Quantity:
5g
25g
100g
13173-09-6
C7H8O
108.14
MFCD00066201
LNLLHUHPGPKRBM-UHFFFAOYSA-N
bicyclo 3.2.0 hept-2-en-6-one,+/--cis-bicyclo 3.2.0 hept-2-en-6-one,cis-bicyclo 3.2.0 hept-2-en-6-one,1s,5s-bicyclo 3.2.0 hept-2-en-6-one,+-1r,5s-cis-bicyclo 3.2.0 hept-2-en-6-one,bicyclo 3,2,0 hept-2-en-6-one,bicyclo 3,2,0 hept-2-ene-6-one,bicyclo 3.2.0 hept-3-en-7-one,z-bicyclo 3.2.0 hepta-2-ene-6-one
297183
bicyclo[3.2.0]hept-3-en-7-one
C1C=CC2C1C(=O)C2
13173-09-6
C7H8O
108.14
MFCD00066201
LNLLHUHPGPKRBM-UHFFFAOYSA-N
bicyclo 3.2.0 hept-2-en-6-one,+/--cis-bicyclo 3.2.0 hept-2-en-6-one,cis-bicyclo 3.2.0 hept-2-en-6-one,1s,5s-bicyclo 3.2.0 hept-2-en-6-one,+-1r,5s-cis-bicyclo 3.2.0 hept-2-en-6-one,bicyclo 3,2,0 hept-2-en-6-one,bicyclo 3,2,0 hept-2-ene-6-one,bicyclo 3.2.0 hept-3-en-7-one,z-bicyclo 3.2.0 hepta-2-ene-6-one
297183
bicyclo[3.2.0]hept-3-en-7-one
C1C=CC2C1C(=O)C2
Applications
Bicyclo[3.2.0]hept-2-en-6-one is widely utilized to study enantioselective Baeyer-Villiger oxidation of (±)-cis-bicyclo(3.2.0)hept-2-en-6-one. It is used in the synthesis of a series of chalcone derivatives by reacting with arylaldehydes. It is also used to analyze the extremophile enzymes (monooxygenase and hydrolases) in microorganisms which are isolated from a deep-water petroleum reservoir and at high temperatures using fluorogenic assays and multibioreactions.

Notes
Air sensitive. Incompatible with strong oxidizing agents.
Quantity 5g
UN Number UN1224
Formula Weight 108.14
Percent Purity 97%
Chemical Name or Material Bicyclo[3.2.0]hept-2-en-6-one

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KETONES, LIQUID, N.O.S.