Learn More
BID Antibody (OTI1G10), Biotin, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP270254B
Catalog No. NBP270254B
Add to Cart
Description
Description
BID Monoclonal specifically detects BID in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
Specifications
BID
Monoclonal
Biotin
PBS with 0.05% Sodium Azide
BID
Full length recombinant protein of human BID (NP_001187) produced in HEK293T cells.
0.1 mL
Primary
Human
IgG2a
Western Blot, Immunohistochemistry
OTI1G10
Western Blot, Immunohistochemistry
apoptic death agonist, BH3 interacting domain death agonist, BH3-interacting domain death agonist, BID isoform ES(1b), BID isoform L(2), BID isoform Si6, desmocollin type 4, FP497, Human BID coding sequence, MGC15319, MGC42355, p22 BID, tbid
Mouse
Affinity Purified
Apoptosis, Cancer, Phospho Specific, Tumor Suppressors
637.0
Store at 4°C in the dark.
Product Suggestions
Product Suggestions