Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BID Mouse anti-Human, Alexa Fluor™ 647, Clone: OTI1G10, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270254AF647

Catalog No. N270254A647


Add to cart

Description

Description

BID Monoclonal antibody specifically detects BID in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

BID
OTI1G10
Western Blot, Immunohistochemistry
Affinity Purified, Conjugated
BID
Full length recombinant protein of human BID (NP_001187) produced in HEK293T cells.
Immunogen affinity purified
Apoptosis, Cancer, Phospho Specific, Tumor Suppressors
Primary
637.0
Immunohistochemistry, Western Blot
Alexa Fluor 647
50mM Sodium Borate with 0.05% Sodium Azide
apoptic death agonist, BH3 interacting domain death agonist, BH3-interacting domain death agonist, BID isoform ES(1b), BID isoform L(2), BID isoform Si6, desmocollin type 4, FP497, Human BID coding sequence, MGC15319, MGC42355, p22 BID
Mouse
IgG2a
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit