Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Biebrich Scarlette, HARLECO™, MilliporeSigma™

Click to view available options
Quantity:
25g
4196-99-0
C22H14N4Na2O7S2
556.475
YCKMEQRBEVVZQF-UHFFFAOYSA-L
Crocein Scarlet
16219040
disodium;2-[2-(2-oxonaphthalen-1-ylidene)hydrazinyl]-5-[(4-sulfonatophenyl)diazenyl]benzenesulfonate
C1=CC=C2C(=C1)C=CC(=O)C2=NNC3=C(C=C(C=C3)N=NC4=CC=C(C=C4)S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+]
4196-99-0
C22H14N4Na2O7S2
556.475
YCKMEQRBEVVZQF-UHFFFAOYSA-L
Crocein Scarlet
16219040
disodium;2-[2-(2-oxonaphthalen-1-ylidene)hydrazinyl]-5-[(4-sulfonatophenyl)diazenyl]benzenesulfonate
C1=CC=C2C(=C1)C=CC(=O)C2=NNC3=C(C=C(C=C3)N=NC4=CC=C(C=C4)S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+]

Biebrich Scarlette; Certified biological stain; Dark red powder; EMD; HARLECO; 25g; Glass bottle; Crocein Scarlet; CAS: 4196-99-0

Biebrich Scarlette; Certified biological stain; Dark red powder; EMD; HARLECO; 25g; Glass bottle; Crocein Scarlet; CAS: 4196-99-0

Physical Form Powder
Grade Certified Biological Stain
Quantity 25g
Chemical Name or Material Biebrich Scarlette, HARLECO™