Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bifemelane hydrochloride, Tocris Bioscience™

Click to view available options
Quantity:
10mg
50mg

Chemical Identifiers

62232-46-6
C18H24ClNO
305.846
MEAHDXWXNNDSAK-UHFFFAOYSA-N
4-(2-Benzylphenoxy)-N-methylbutylamine hydrochloride
6917789

MAO-A and MAO-B inhibitor

Antidepressant MAO inhibitor. Reverses catalepsy induced by tetrabenazine in mice and increases locomotor activity in MPTP-treated marmosets.
Quantity 10mg
SMILES CNCCCCOC1=CC=CC=C1CC2=CC=CC=C2.Cl
IUPAC Name 4-(2-benzylphenoxy)-N-methylbutan-1-amine;hydrochloride
Formula Weight 305.84
Chemical Name or Material Bifemelane hydrochloride
Grade >99%

Recommended Storage : Desiccate at Room Temperature