Learn More
Enzo Life SciencesSupplier Diversity Partner Big Endothelin-1 (human), ELISA kit (1x96wells)  DFS Item
Manufacturer:  Enzo Life SciencesSupplier Diversity Partner ADI900022
The Big Endothelin-1 (human), EIA kit is a colorimetric immunometric enzyme immunoassay kit with results overnight + 1 hour. Alternative Name: Big ET-1. Sensitivity: 0.23 pg/ml (range 0.78 - 100 pg/ml). Assay Time: Overnight + 1 hour. Applications: ELISA, Colorimetric detection. Species Reactivity: Human. Contents: Microtiter plate, Labeled antibody concentrate, Antibody diluent, Assay buffer, Wash buffer concentrate, Standard, TMB Substrate, Stop solution
Catalog No. 50-658-107