Learn More
Applied Biosystems™ BigDye™ Terminator v3.1 Cycle Sequencing Kit Non-distribution product as customer accommodation.
The BigDye™Terminator v3.1 Cycle Sequencing Kit's robust, highly flexible chemistry
$10540.00 - $48500.00
Products 2
Catalog Number Mfr. No. Description For Use With (Application) For Use With (Equipment) Includes Price Quantity & Availability  
Catalog Number Mfr. No. Description For Use With (Application) For Use With (Equipment) Includes Price Quantity & Availability  
43-374-56 Non-distribution product as customer accommodation. View Documents Applied Biosystems™
4337456
Cycle Sequencing Kit, 1000 rxns, Includes 10 Tubes of BigDye™ Terminator v3.1 Ready Reaction Mix, M13 (-21) Primer, pGEM™-3Zf(+) double-stranded DNA Control Template, 1 x 12mL tube of 5X Sequencing Buffer (may be stored at 4°C) Ideal for De Novo Sequencing, Resequencing and Finishing with PCR Product, Plasmid, Fosmid and BAC templates 310 Genetic Analyzer, 3130 Genetic Analyzer, 3130xl Genetic Analyzer, 3500 Genetic Analyzer, 3500xL Genetic Analyzer, 3730 DNA Analyzer, 3730xl DNA Analyzer, GeneAmp™ 9700, Veriti™ Thermal Cycler 10 Tubes of BigDye™ Terminator v3.1 Ready Reaction Mix, M13 (-21) Primer, pGEM™-3Zf(+) double-stranded DNA Control Template, 1 x 12mL tube of 5X Sequencing Buffer (may be stored at 4°C) Each for $10,540.00
Add to cart
43-374-57 Non-distribution product as customer accommodation. View Documents Applied Biosystems™
4337457
Cycle Sequencing Kit, 5000 rxns, Includes 2 x 20mL tubes of BigDye™ Terminator v3.1 Ready Reaction Mix, 2 tubes M13 (-21) Primer, 2 tubes pGEM Control DNA, 2 x 28mL tubes of 5X Sequencing Buffer (may be stored at 4°C) Ideal for De Novo Sequencing, Resequencing and Finishing with PCR Product, Plasmid, Fosmid and BAC templates 310 Genetic Analyzer, 3130 Genetic Analyzer, 3130xl Genetic Analyzer, 3500 Genetic Analyzer, 3500xL Genetic Analyzer, 3730 DNA Analyzer, 3730xl DNA Analyzer, GeneAmp™ 9700, Veriti™ Thermal Cycler 2 x 20mL tubes of BigDye™ Terminator v3.1 Ready Reaction Mix, 2 tubes M13 (-21) Primer, 2 tubes pGEM Control DNA, 2 x 28mL tubes of 5X Sequencing Buffer (may be stored at 4°C) Each for $48,500.00
Add to cart
Description
Description
  • Improve the quality of your results for a wide range of sequencing applications
  • Optimized for long read lengths
  • Better dye mobility characteristics
  • Improved performance reading through GT rich regions
  • Get longer, higher-quality reads with more uniform peak heights and optimal signal balance
  • Enhance your productivity and reduce costs