Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BIIB 021, Tocris Bioscience™

Catalog Number 460850
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
848695-25-0
C14H15ClN6O
318.765
QULDDKSCVCJTPV-UHFFFAOYSA-N
16736529
6-chloro-9-[(4-methoxy-3,5-dimethylpyridin-2-yl)methyl]purin-2-amine
CC1=CN=C(C(=C1OC)C)CN2C=NC3=C2N=C(N=C3Cl)N
This item is not returnable. View return policy
848695-25-0
C14H15ClN6O
318.765
QULDDKSCVCJTPV-UHFFFAOYSA-N
16736529
6-chloro-9-[(4-methoxy-3,5-dimethylpyridin-2-yl)methyl]purin-2-amine
CC1=CN=C(C(=C1OC)C)CN2C=NC3=C2N=C(N=C3Cl)N

Selective Hsp90 inhibitor

Selective, Hsp90 competitive inhibitor (Ki = 1.7 nM). Displays no significant activity at a range of ATP-binding kinases or Na+K+ ATPase. Induces degradation of HER-2 in vitro (EC50 = 38 nM in MCF-7 cells). Inhibits growth and promotes cell death in a variety of human tumor cells. Orally bioavailable.
Quantity 50 mg
Synonym biib021, 6-chloro-9-4-methoxy-3,5-dimethylpyridin-2-yl methyl-9h-purin-2-amine, biib 021, biib-021, unii-851b9fq7q0, 6-chloro-9-4-methoxy-3,5-dimethylpyridin-2-yl methyl purin-2-amine, 6-chloro-9-4-methoxy-3,5-dimethyl-2-pyridinyl methyl-9h-purin-2-amine, 3qdd
Formula Weight 318.76
Percent Purity >98%
Chemical Name or Material BIIB 021
Recommended Storage Store at -20°C
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit