Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BIIE 0246, Tocris Bioscience™

Click to view available options
Quantity:
1mg
10mg
This item is not returnable. View return policy
246146-55-4
C49H57N11O6
896.066
RSJAXPUYVJKAAA-JPGJPTAESA-N
d06gfz, cyclopentaneacetamide, n-1s-4-aminoiminomethyl amino-1-2-3,5-dioxo-1,2-diphenyl-1,2,4-triazolidin-4-yl ethyl amino carbonyl butyl-1-2-4-6,11-dihydro-6-oxo-5h-dibenz b,e azepin-11-yl-1-piperazinyl-2-oxoethyl, 2s-5-diaminomethylideneamino-n-2-3,5-dioxo-1,2-di phenyl-1,2,4-triazolidin-4-yl ethyl-2-2-1-2-oxo-2-4-6-oxo-5,11-dihydrobenzo c 2 benzazepin-11-yl piperazin-1-yl ethyl cyclopentyl acetyl amino pentanamide, 2s-5-carbamimidamido-n-2-3,5-dioxo-1,2-diphenyl-1,2,4-triazolidin-4-yl ethyl-2-2-1-2-oxo-2-4-10-oxo-9-azatricyclo 9.4.0.0 3 ,? pentadeca-1 15 ,3,5,7,11,13-hexaen-2-yl piperazin-1-yl ethyl cyclopentyl acetamido pentanamide, n-1s-4-amino-iminomethyl amino-1-2-3,5-dioxo-1,2-diphenyl-1,2,4-triazolidin-4-yl ethyl-amino carbonyl butyl-1-2-4-6,11-dihydro-6-oxo-5h-dibenz b,e azepin-11-yl-1-piperazinyl-2-oxoethyl-cyclopentaneacetamide, n-1s-4-aminoiminomethyl amino-1-2-3,5-dioxo-1,2-diphenyl-1,2,4-triazolidin-4-yl ethyl amino carbonyl butyl-1-2-4-6,11-dihydro-6-oxo-5h-dibenz b,e azepin-11-yl-1-piperazinyl-2-oxoethyl-cyclopentaneacetamide
9811493
(2S)-5-(diaminomethylideneamino)-N-[2-(3,5-dioxo-1,2-diphenyl-1,2,4-triazolidin-4-yl)ethyl]-2-[[2-[1-[2-oxo-2-[4-(6-oxo-5,11-dihydrobenzo[c][1]benzazepin-11-yl)piperazin-1-yl]ethyl]cyclopentyl]acetyl]amino]pentanamide
C1CCC(C1)(CC(=O)NC(CCCN=C(N)N)C(=O)NCCN2C(=O)N(N(C2=O)C3=CC=CC=C3)C4=CC=CC=C4)CC(=O)N5CCN(CC5)C6C7=CC=CC=C7C(=O)NC8=CC=CC=C68
This item is not returnable. View return policy
246146-55-4
C49H57N11O6
896.066
RSJAXPUYVJKAAA-JPGJPTAESA-N
d06gfz, cyclopentaneacetamide, n-1s-4-aminoiminomethyl amino-1-2-3,5-dioxo-1,2-diphenyl-1,2,4-triazolidin-4-yl ethyl amino carbonyl butyl-1-2-4-6,11-dihydro-6-oxo-5h-dibenz b,e azepin-11-yl-1-piperazinyl-2-oxoethyl, 2s-5-diaminomethylideneamino-n-2-3,5-dioxo-1,2-di phenyl-1,2,4-triazolidin-4-yl ethyl-2-2-1-2-oxo-2-4-6-oxo-5,11-dihydrobenzo c 2 benzazepin-11-yl piperazin-1-yl ethyl cyclopentyl acetyl amino pentanamide, 2s-5-carbamimidamido-n-2-3,5-dioxo-1,2-diphenyl-1,2,4-triazolidin-4-yl ethyl-2-2-1-2-oxo-2-4-10-oxo-9-azatricyclo 9.4.0.0 3 ,? pentadeca-1 15 ,3,5,7,11,13-hexaen-2-yl piperazin-1-yl ethyl cyclopentyl acetamido pentanamide, n-1s-4-amino-iminomethyl amino-1-2-3,5-dioxo-1,2-diphenyl-1,2,4-triazolidin-4-yl ethyl-amino carbonyl butyl-1-2-4-6,11-dihydro-6-oxo-5h-dibenz b,e azepin-11-yl-1-piperazinyl-2-oxoethyl-cyclopentaneacetamide, n-1s-4-aminoiminomethyl amino-1-2-3,5-dioxo-1,2-diphenyl-1,2,4-triazolidin-4-yl ethyl amino carbonyl butyl-1-2-4-6,11-dihydro-6-oxo-5h-dibenz b,e azepin-11-yl-1-piperazinyl-2-oxoethyl-cyclopentaneacetamide
9811493
(2S)-5-(diaminomethylideneamino)-N-[2-(3,5-dioxo-1,2-diphenyl-1,2,4-triazolidin-4-yl)ethyl]-2-[[2-[1-[2-oxo-2-[4-(6-oxo-5,11-dihydrobenzo[c][1]benzazepin-11-yl)piperazin-1-yl]ethyl]cyclopentyl]acetyl]amino]pentanamide
C1CCC(C1)(CC(=O)NC(CCCN=C(N)N)C(=O)NCCN2C(=O)N(N(C2=O)C3=CC=CC=C3)C4=CC=CC=C4)CC(=O)N5CCN(CC5)C6C7=CC=CC=C7C(=O)NC8=CC=CC=C68

Potent, selective non-peptide NPY Y2 receptor antagonist

Potent, selective and competitive non-peptide antagonist for the neuropeptide Y Y2 receptor (IC50 = 15 nM). Displays > 650-fold selectivity over Y1, Y4 and Y5 receptors. Active in vivo.
Quantity 10mg
Formula Weight 896.06
Percent Purity >98%
Chemical Name or Material BIIE 0246

Recommended Storage : Store at +4°C