Learn More
BioVision IncSupplier Diversity Partner Total Bile Acids (TBA) Assay Kit (Colorimetric): Total Bile Acids (TBA) Assay Kit (Colorimetric): A fast, sensitive, easy to use colorimetric assay to measure total bile acids in various biological samples. 100 assays DFS Item
Manufacturer:  BioVision IncSupplier Diversity Partner K209100
Total Bile Acids (TBA) Assay Kit (Colorimetric): Bile Acids (BA) make 67% of the total composition of Bile. Bile acids are critical due to their ability to solubilize lipids by forming micelles with cholesterol, and fatty acids. BioVision's Total Bile Acid Assay Kit provides a simple, sensitive, and high-throughput adaptable approach to detect physiological concentration of total bile acids in a variety of biological fluids. The principle of the assay is based on an enzymatic cycling method in the presence of NADH and a chromophore. The reduction of the chromophore produces a stable colorimetric product the absorbance of which can be followed kinetically at 405 nm. This absorbance is directly proportional to the amount of TBA in the sample. Our assay is very specific and sensitive. Other metabolites found in biofluids do not interfere with the assay. The assay can detect as little as 1 ¿M of Bile Acids in a variety of samples. BioVision charges an Ice pack fee for each order.
Catalog No. 50-111-9621