Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bile Salts, Spectrum™ Chemical
Click to view available options
Quantity:
25 g
100 g
500 g
5 kg
Packaging:
Poly Bottle
Poly Pail
Description
Spectrum™ Chemical Bile Salts are also known as bile acids. They are steroidal acids that a thick yellow fluid made by the liver. It helps digest the fat in your diet. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.
Specifications
CAS Max % | 100% |
Packaging | Poly Bottle |
Quantity | 100 g |
Grade | Reagent |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction