Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Biliverdin hydrochloride, Thermo Scientific Chemicals

Catalog Number p-7043046
Click to view available options
Quantity:
100 mg
250 mg
55482-27-4
C33H36Cl2N4O6
655.573
MFCD08669554
GNLHFBAZSKVVCQ-VTURHDOZSA-N
biliverdin hydrochloride
132991794
3-[(2Z)-2-[[3-(2-carboxyethyl)-5-[(E)-(3-ethenyl-4-methyl-5-oxopyrrol-2-ylidene)methyl]-4-methyl-1H-pyrrol-2-yl]methylidene]-5-[(4-ethenyl-3-methyl-5-oxopyrrol-2-yl)methylidene]-4-methylpyrrol-3-yl]propanoic acid;dihydrochloride
CC1=C(C(=CC2=C(C(=C(N2)C=C3C(=C(C(=O)N3)C)C=C)C)CCC(=O)O)NC1=CC4=NC(=O)C(=C4C)C=C)CCC(=O)O.Cl.Cl
55482-27-4
C33H36Cl2N4O6
655.573
MFCD08669554
GNLHFBAZSKVVCQ-VTURHDOZSA-N
biliverdin hydrochloride
132991794
3-[(2Z)-2-[[3-(2-carboxyethyl)-5-[(E)-(3-ethenyl-4-methyl-5-oxopyrrol-2-ylidene)methyl]-4-methyl-1H-pyrrol-2-yl]methylidene]-5-[(4-ethenyl-3-methyl-5-oxopyrrol-2-yl)methylidene]-4-methylpyrrol-3-yl]propanoic acid;dihydrochloride
CC1=C(C(=CC2=C(C(=C(N2)C=C3C(=C(C(=O)N3)C)C=C)C)CCC(=O)O)NC1=CC4=NC(=O)C(=C4C)C=C)CCC(=O)O.Cl.Cl

Biliverdin dihydrochloride was found responsible for cytoprotection. It was also found to induce tolerance to cardiac allografts. Biliverdin hydrochloride may regulate the cellular heme degradation process by occupying the heme binding site on heme oxygenase which would prevent access of the substrate to the catalytic site of the enzyme.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Biliverdin dihydrochloride was found responsible for cytoprotection. It was also found to induce tolerance to cardiac allografts. Biliverdin hydrochloride may regulate the cellular heme degradation process by occupying the heme binding site on heme oxygenase which would prevent access of the substrate to the catalytic site of the enzyme.

Solubility
Soluble in DMSO, DMF, and basic aqueous solutions

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
TRUSTED_SUSTAINABILITY
Color Black to Green
Quantity 100 mg
Sensitivity Light sensitive
Solubility Information Soluble in DMSO, DMF, and basic aqueous solutions
Formula Weight 619.12
Physical Form Powder
Chemical Name or Material Biliverdin hydrochloride
RTECSNumber DU3033000
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit