Learn More
Tocris Bioscience™ BIM 23056 Available on GSA/VA Contract for Federal Government customers only.
sst receptor ligand
Manufacturer:  Tocris Bioscience™ 1844/1
Catalog No. 18-441-R
Add to cart
This item is not returnable. View return policy
Description
Description
Somatostatin receptor ligand (Ki values are 142, > 1000, 10.8, 16.6 and 5.7 nM for human cloned sst1 - 5 receptors respectively). Behaves as an antagonist on SRIF14-induced [35S]-GTPγS binding at sst3 and sst5 receptors (PKB values are 6.33 and 5.84 respectively).
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.