Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bim Mouse anti-Human, Janelia Fluor 646, Clone: 1C2H4, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP175511JF646

Catalog No. NBP75511646


Add to cart

Description

Description

Bim Monoclonal antibody specifically detects Bim in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

Bim
1C2H4
Immunocytochemistry/Immunofluorescence
Affinity Purified
BCL2L11
Antibody was raised against a recombinant protein corresponding to amino acids 2 to 198 of human Bim.
Protein A purified
Apoptosis, Cancer, Immunology, Innate Immunity, Tumor Suppressors
Primary
10018
Immunocytochemistry, Immunofluorescence
Janelia Fluor 646
50mM Sodium Borate with 0.05% Sodium Azide
BAM, bcl-2 interacting mediator of cell death, bcl-2 interacting protein Bim, Bcl2-interacting mediator of cell death, bcl2-L-11, BCL2-like 11 (apoptosis facilitator), bcl-2-like protein 11, bcl-2-related ovarian death agonist, BIM-alpha6, BIM-beta6, BIM-beta7, BIMBOD, BimEL, BimL
Mouse
IgG1
0.1mL
Store at 4°C in the dark.
Monoclonal
Human, Mouse, Rat
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit