Alfa Aesar™ Bindone, 98+%

Manufacturer: Alfa Aesar™ B2455706

Quantity 5g

 View more versions of this product

Catalog No. AAB2455706

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 1707-95-5
    Molecular Formula C18H10O3
    Formula Weight 274.28
    MDL Number MFCD00021233
    Beilstein 1884961
    Synonym 2-[1-(3-Oxoindanylidene)]-1,3-indanedione
    Chemical Name or Material Bindone

    Specifications

    Formula Weight 274.28
    Molecular Formula C18H10O3
    MDL Number MFCD00021233
    Beilstein 1884961
    Melting Point 205° to 208°C
    Synonym 2-[1-(3-Oxoindanylidene)]-1,3-indanedione
    Quantity 5g
    RTECS Number NK6050000
    CAS 1707-95-5
    Chemical Name or Material Bindone
    Assay ≥98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit