Learn More
Bio/Data™ Beta Pak (ADP, Collagen, Ristocetin)
Formulated to differentiate platelet function disorders and von Willebrand Syndrome
Manufacturer:  Bio/Data™ 101580
Catalog No. 22-515-232
Add to cart
This item is not returnable. View return policy
Description
Description
  • Contains 0.5mL each of ADP, Soluble Type I Collagen and Ristocetin
  • All reagents are Lyophilized to enable long term storage and stability
Specifications
Specifications
Routine in-vitro studies for differential diagnosis of platelet disorders and von Willebrand syndrome
4M ADP (Adenosine-5'-Diphosphate), 1.9mg/mL soluble Collagen and 15mg/mL Ristocetin
BETA/Pak™ kit
Lyophilized
1 x 0.5mL