Learn More
Bio/Data Lupus Anticoagulant Confirmation Reagent (LA-CR)
Designed to Perform platelet neutralization procedure (PNP)
Manufacturer: Bio/Data 102516
This item is not returnable.
View return policy
Description
- Evaluate prolonged Activated Partial Thromboplastin Time (APTT) which is greater than 5 seconds above the upper limit of the normal range
- Lyophilized preparation of phosphatidyl enhanced platelet phospholipid
- Lyophilization enables long term storage and stability
Specifications
Lyophilized | |
To differentiate lupus anticoagulant | |
Platelet Extract Reagent | |
5 x 1mL |