Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Bio/Data™ Lyophilized Platelets for von Willebrand Factor Quantitation
For use in quantification of von Willebrand factor in plasma
Manufacturer:  Bio/Data™ 101258
Catalog No. 22-515-249
This item is not returnable. View return policy
Description
Description
  • Standardized and fixed platelet suspension derived from human blood
  • Tris buffered saline
  • Lyophilization enables long term storage and stability
  • Approximate platelet count of 200000/µL
  • Quantity: 1 x 10mL
Specifications
Specifications
Tris Buffered Saline Diluent
Each
Lyophilized Pellets