Learn More
Bio Plas™ Uni-Capsettes™ Process/Embedding Cassettes
Provides a modular and organized means for safe and consistent processing, embedding, sectioning, and storage of specimen samples
Supplier: Bio Plas™ 6058
This item is not returnable.
View return policy
Description
- Detachable plastic lids
- Heavy wall construction will not float or open during processing
- Engineered to eliminate“chatter”during sectioning
- Strategically designed slots allow for efficient solvent flow and drainage
- Special acetyl copolymer plastic resists heat, solvents, decacifying solutions, and problems with microwave processing
- Fit standard stainless steel and disposable plastic base molds
- Accepted by all current automatic cassette labelers
Specifications
Separate Lid | |
Gold | |
Acetyl Copolymer Plastic | |
Tissue Cassette |