Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio Plas™ Uni-Flex™ Safety Caps

Catalog No. p-4422869
Click to view available options
Color:
Gray
Green
Lavender
Natural
Red
Yellow
For Use With (Equipment):
12 mm Culture or 13 mm Blood Collection Tubes
13 mm Culture Tubes
13 mm Culture and Blood Collection Tubes
16 mm Culture and Blood Collection Tubes

Protects laboratory staff from infectious aerosols

  • Liquid-tight seal ensures sample integrity and prevent spillage of serum samples
  • Designed to hold under centrifugation, agitation, refrigeration and freezing
  • Stretchable, unique, over-locking design to prevent evaporation
  • One-hand application and removal
  • Trace-metal free material

Specifications

Includes Liquid-tight seal
Product Type Safety Cap
Material Polyethylene
Color Green
For Use With (Equipment) 13 mm Culture Tubes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.