Bio-Serv™ AIN-93M Rodent Diet Available on GSA/VA Contract for Federal Government customers only.

Purified diet formulated to meet the nutritional requirements of mature rodents

Manufacturer: BIO SERV F3155

 View more versions of this product

Catalog No. 14-726-617

  Check Availability
Add to Cart

    Specifications

    Quantity 5000g
    Species Rodent
    Components Corn Starch, Casein, Maltodextrin, Sucrose, Soybean Oil, Cellulose, Mineral Mix, Vitamin Mix, L-Cystine, Choline Bitartrate, tBHQ
    Diameter (English) 0.5 in.
    Product Type Rodent diet
    Diameter (Metric) 1.27cm
    Shelf Life 6 Months
    Format Pellet
    For Use With (Application) Used and published for research objectives
    Particle Shape Pellet

    • Widely used and published
    • Phytoestrogen free
    • Chlorophyll free—does not interfere with imaging studies
    • Nutritionally assayed

    • Ingredients: Corn Starch, Casein, Maltodextrin, Sucrose, Cellulose, Soybean Oil, Mineral Mix, Vitamin Mix, Choline Bitartrate, L-Cystine, tBHQ

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit