Bio-Serv™ Bacon Yummies™ Enrichment Treats, Certified Available on GSA/VA Contract for Federal Government customers only.

Nutritionally complete grain-based treat with a highly palatable bacon flavor

Manufacturer: BIO SERV F05778

 View more versions of this product

Catalog No. 14-726-580

  Check Availability
Add to Cart

    Specifications

    Flavor Bacon
    Storage Requirements Cool dry place
    Format Pellet
    Quantity 20,000/bag
    Components Ground Corn, Banana Flakes, Dehulled Soybean Meal, Corn Gluten Meal, Ground Wheat, Corn Gluten Feed, Fish Meal, Dehydrated Alfalfa Meal, Casein, Dried Whey, Sucrose, Fructose, Dextrose, Dried Beet Pulp, Soybean Oil, Porcine Animal Fat (preserved with BHA), Dried Brewers Yeast, Mineral Mix, Vitamin Mix, Bacon Flavor, Magnesium Stearate, Calcium Silicate, DL-Methionine, Choline Chloride, Sodium Propionate, Ascorbic Acid, Kaolin
    Calories 3.37
    Product Type Bacon Yummies
    Shelf Life 6 Months
    Particle Shape Pellet

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit