Bio-Serv™ Bimple Bones Animal Diet, Certified

Safe and easily digestible edible bones

Manufacturer: BIO SERV K3362

 View more versions of this product

Catalog No. 14-726-710

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    Flavor Peanut butter
    Storage Requirements Cool dry place
    For Use With (Application) Tartar control
    Quantity Each
    Components Modified Corn Starch, Water, Glycerin, Sorbitol, Cellulose, Dried Yeast Hydrolysate, Lecithin, Calcium Stearate, Sorbic Acid, Food Dye, Natural Flavor
    Product Type Bimple bones
    Shelf Life 1 Year
    Particle Shape Bone

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit