Bio-Serv™ Crumble Disks™ Animal Diet Available on GSA/VA Contract for Federal Government customers only.

Manufacturer: BIO SERV F05049CD

 View more versions of this product

Catalog No. 14-726-724

  Check Availability
Add to Cart

    Specifications

    Flavor Mixed Fruit
    Storage Requirements Cool dry place
    Format Crumbles
    For Use With (Application) GLP and TOX studies
    Quantity 72/box
    Product Type Crumble disks
    Shelf Life 6 Months
    Particle Shape Crumble Disks

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit