Learn More
Bio-Serv™ Dustless Precision Pellets™ Grain-Based Rodent Diet Available on GSA/VA Contract for Federal Government customers only.
Precise size and weight pellets generally used in nutritional and behavioral studies
Manufacturer:  BIO SERV F0170
 View more versions of this product
Catalog No. 14-726-584
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Pellet
Rodent
3.35
Rodent diet
Cool dry place
Nutritional and behavioral studies
0.3g
Ground Corn, Banana Flakes, Dehulled Soybean Meal, Corn Gluten Meal, Ground Wheat, Corn Gluten Feed, Fish Meal, Dehydrated Alfalfa Meal, Casein, Dried Whey, Sucrose, Fructose, Dextrose, Dried Beet Pulp, Soybean Oil, Porcine Animal Fat (preserved with BHA), Dried Brewers Yeast, Mineral Mix, Vitamin Mix, Magnesium Stearate, DL-Methionine, Choline Chloride, Sodium Propionate, Ascorbic Acid, Kaolin
0.3 in.
0.076cm
Pellet
Safety and Handling
ShelfLife : 1 Year