Learn More
Bio-Serv™ Dustless Precision Pellets™ Purified Rodent Diet Available on GSA/VA Contract for Federal Government customers only.
Precise size and weight pellets generally used in nutritional and behavioral studies
Manufacturer:  BIO SERV F0077
 View more versions of this product
Catalog No. 14-726-593
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Pellet
Rodent
3.6
Rodent diet
No Added Flavor
Nutritional and behavioral studies
0.75g
Sucrose. Dextrose, Casein, Corn Oil, Mineral Mix, Cellulose, Corn Syrup, Calcium Silicate, Vitamin Mix, Magnesium Stearate, Choline Bitartrate, DL-Methionine, L-Cystine, Ascorbic Acid, Vitamin E Acetate, tBHQ
0.38 in.
0.097cm
Cool dry place
Safety and Handling
ShelfLife : 1 Year