Bio-Serv™ Dustless Precision Pellets™ for Rodents, Purified Available on GSA/VA Contract for Federal Government customers only.

This nutritional treat is used in various rodent studies

Manufacturer: BIO SERV F0076

 View all options for this product

Catalog No. 14-726-754

  Check Availability
Add to Cart


    Quantity 0.5g
    Species Rodent
    Components Sucrose, Dextrose, Casein, Corn Oil, Mineral Mix, Cellulose, Corn Syrup, Calcium Silicate, Vitamin Mix, Magnesium Stearate, Choline Bitartrate, DL-Methionine, L-Cystine, Ascorbic Acid, Vitamin E Acetate, tBHQ
    Calories 3.6
    Diameter (English) 0.36 in.
    Product Type Rodent diet
    Diameter (Metric) 0.9cm
    Shelf Life 1 Year
    Flavor Custom
    Storage Requirements Cool dry place
    Format Pellet
    For Use With (Application) Used in nutritional and behavioral studies
    Particle Shape Pellet

    • Precise size and weight pellets generally used in nutritional and behavioral studies
    • Dustless and fracture free
    • Superior performance in automatic feeders
    • Nutritionally complete purified formula
    • Every batch tested for hardness, weight and height
    • Particle shape: pellet
    • Nutritionally assayed

    Generally used in nutritional and behavioral studies

  • Description & Specifications

Related Products

Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit