Bio-Serv™ Fruit and Nut Foraging Mix, Certified Available on GSA/VA Contract for Federal Government customers only.

Natural foraging mixture

Manufacturer: BIO SERV F3084

 View more versions of this product

Catalog No. 14-726-771

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »


    Specifications

    Flavor Banana
    Storage Requirements Cool dry place
    For Use With (Application) Use with tuff turf foraging board or scatter into bedding
    Quantity 10000g
    Components Peanuts, Sunflower Seeds, Banana Chips and Raisins
    Calories 5.26
    Product Type Fruit and nut foraging mix
    Shelf Life 6 Months
    Particle Shape Dry Mix

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit