Learn More
Bio-Serv™ Holder for Pellet Feeding Tubes Available on GSA/VA Contract for Federal Government customers only.
Bottom tray catches any reclaimable pellet fines
Manufacturer:  BIO SERV 90055
 View more versions of this product
Catalog No. 14-726-521
Due to product restrictions, please to purchase or view availability for this product.
Specifications Description & Specifications
Specifications
Designed to hang over lip of cage
Yes
1.14cm
1.37 in.
Narrow Opening Tube holder
4.5 in.
Stainless Steel
0.348cm