Learn More
Bio-Serv™ Kahn-Pregnant/Weanling Rodent Liquid Diet Available on GSA/VA Contract for Federal Government customers only.
Versatile liquid format allows the addition of alcohol, medications, or test compounds
Manufacturer:  BIO SERV F0504
 View more versions of this product
Catalog No. 14-726-628
  Check Availability
Add to cart
Requires : Blender required for mixing.
Specifications Description & Specifications
Specifications
Powder
Rodent
1000
Powder
4L
Corn syrup solids, Casein, Olive Oil, Maltodextrin, Mineral Mix, Corn Oil, Vitamin Mix, Cellulose, Safflower Oil, Suspending Aid, Choline Dihydrogen Citrate, L-Cystine, DL-Methionine
Rodent liquid diet- Ethanol
Ideal for post operative recovery
Safety and Handling
ShelfLife : 6 Months