Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio-Serv™ Liquid Diet Feeding Tubes Available on GSA/VA Contract for Federal Government customers only.

Designed specifically for liquid diets—eliminates clogging that occurs with sipper tubes

Manufacturer:  BIO SERV 9007

 View more versions of this product

Catalog No. 14-726-525


  Check Availability
Add to cart

Requires : For use with stainless steel tube holders (Mfr. Nos. 9015 and 9016)

Specifications Description & Specifications

Specifications

For liquid diet feeding
Yes
0.15L
17.8cm
1.62 in.
Liquid diet feeding tube
0.039 gal.
7 in.
Borosilicate Glass
0.41cm
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit