Bio-Serv™ Low Fat Lieber-DeCarli '83, '89 Rodent Liquid Diet, Shake and Pour Available on GSA/VA Contract for Federal Government customers only.

No blender needed—just add water, shake, and pour

Manufacturer: BIO SERV F1340SP

 View more versions of this product

Catalog No. 14-726-633

  Check Availability
Add to Cart

    Specifications

    Storage Requirements Cool dry place
    Format Powder
    For Use With (Application) Ideal for post operative recovery
    Quantity 4L
    Species Rodent
    Components Corn syrup solids, Casein, Cellulose, Corn Oil, Maltodextrin, Mineral Mix, Olive Oil, Vitamin Mix, Suspending aids, Safflower Oil, L-Cystine, Choline Bitartrate, DL-Methionine
    Calories 1000
    Product Type Rodent liquid diet- Control
    Shelf Life 6 Months
    Particle Shape Powder

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit