Bio-Serv™ Mouse Igloo™ Rodent Enrichment Device, Certified Available on GSA/VA Contract for Federal Government customers only.

Provides shelter and a secluded place to build a nest

Manufacturer: BIO SERV K3327

 View more versions of this product

Catalog No. 14-726-552

  Check Availability
Add to Cart

    Specifications

    Autoclavable Yes
    Color Red
    For Use With (Application) Provide shelter and a secluded place to build a nest, great addition to breeding programs
    Height (English) 2.25 in.
    Height (Metric) 5.7cm
    Material Polycarbonate
    Product Type Mouse igloo

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit