Bio-Serv™ Pellet Feeding Tubes Available on GSA/VA Contract for Federal Government customers only.
Easy, accurate and convenient method of measuring food intake
Manufacturer: BIO SERV 9004
 View more versions of this product
Catalog No. 14-726-526
    N/A
Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 
    Specifications
    Autoclavable Yes
    Height (English) 5.25 in.
    Height (Metric) 1.33cm
    Material Borosilicate Glass
    For Use With (Application) Pellet feeding of Rat
    Product Type Narrow opening Pellet feeding tube
  • Description & Specifications
Related Products
Product Content Feedback