Bio-Serv™ Pellet Feeding Tubes Available on GSA/VA Contract for Federal Government customers only.
Easy, accurate and convenient method of measuring food intake
Manufacturer: BIO SERV 90045
 View more versions of this product
Catalog No. 14-726-520
    N/A
Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 
    Specifications
    Material Borosilicate Glass
    Autoclavable Yes
    For Use With (Application) Pellet feeding of Mice
    Product Type Narrow opening Pellet feeding tube
    Height (English) 3.5 in.
    Height (Metric) 0.89cm
    • Narrow opening allows access to pellet without removal from the tube
    • Made of borosilicate glass
    • Autoclavable
    • Cagewasher safe
  • Description & Specifications
Related Products
Product Content Feedback