Bio-Serv™ PRANG™, Oral Rehydrator Powder

Excellent for crashing, recovering and debilitation conditions

Manufacturer: BIO SERV F2351SGAL

 View more versions of this product

Catalog No. 14-726-436

  Check Availability
Add to Cart

    Specifications

    Flavor Strawberry
    Storage Requirements Cool dry place
    Format Powder
    For Use With (Application) Crashing, recovering, debilitating
    Quantity 100g
    Components Electrolyte, Vitamin C, Mineral, Dextrose, Sodium Chloride, Sodium Bicarbonate, Citric Acid, Potassium Chloride, Ascorbic Acid, Acesulfame Potassium, Natural and Artificial Flavors, Food Dye
    Calories 19.88
    Product Type Oral rehydrator
    Shelf Life 1 Year
    Particle Shape Powder

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit