Bio-Serv™ Pregnant/Weanling, High Protein Lieber-DeCarli '82 Rodent Liquid Diet Available on GSA/VA Contract for Federal Government customers only.

For convenient measurement of food intake or study of nutritional manipulation

Manufacturer: BIO SERV F1265

 View more versions of this product

Catalog No. 14-726-636

  Check Availability
Add to Cart

Required Accessories : Blender required for mixing.

    Specifications

    Format Liquid
    For Use With (Application) Ideal for post operative recovery
    Quantity 4L
    Species Rodent
    Components Corn syrup solids, Casein, Olive Oil, Maltodextrin, Mineral Mix, Corn Oil, Vitamin Mix, Cellulose, Safflower Oil, Suspending Aid, Choline Dihydrogen Citrate, L-Cystine, DL-Methionine
    Calories 1000
    Product Type Rodent liquid diet- Ethanol
    Shelf Life 6 Months
    Particle Shape Liquid

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit