Bio-Serv™ PRIMA-Burger™ Grain-Based Supplement Powder

Adds variety to enrichment programs

Manufacturer: BIO SERV F2212F

 View more versions of this product

Catalog No. 14-726-443

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    Flavor Peanut Butter and Molasses Blend
    Storage Requirements 6 months, refrigerated
    For Use With (Application) Ideal diet to deliver and mask medications
    Quantity 5000g
    Components Ground Corn, Soybean Meal, Corn Gluten Meal, Ground Wheat, Fish Meal, Dehydrated Alfalfa Meal, Casein, Dried Whey, Fructose, Molasses, Beet Pulp, Soybean Oil, Corn Oil, Porcine Animal Fat (preserved with BHA), Peanut Butter, Mineral Mix, Vitamin Mix, Ascorbic Acid
    Calories 4.03
    Product Type Burger
    Shelf Life 6 Months
    Particle Shape Powder

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit