Thermo Scientific™ BioBasic™ 18 LC Columns Available on GSA/VA Contract for Federal Government customers only.

Separate small to medium peptides with these HPLC columns.

Manufacturer: Thermo Scientific™ 72105051030

 View more versions of this product

Catalog No. 72105051030

  Check Availability
Add to Cart

Disclaimers:

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.


    Specifications

    Column Format Analytical Column
    Diameter (Metric) 1mm
    Column Type Reversed Phase
    Max. Pressure 400bar
    Packing Material Spherical, fully porous ultrapure silica
    Particle Size 5µm
    Temperature 60°C
    Pore Size 300Å
    pH 2 to 9
    Stationary Phase C18
    Surface Area 100m²/g
    Type Analytical Column
    Length (Metric) 50mm
    Carbon Load 9%
    USP Type L1
    Endcapped Yes

    Separate small to medium peptides with Thermo Scientific™ BioBasic™ 18 LC Columns.

    • 300Å pore size for maximum performance with biomolecules
    • High peak capacity stationary phase
    • Outstanding reproducibility, efficiency and column lifetime
    • Excellent for LC/MS separations

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit