Learn More
Thermo Scientific™ BioBasic™ 18 Nanobore HPLC Columns
At flow rates of nL/min versus mL/min, BioBasic 18 Nanobore HPLC Columns offer greater signal to noise ratio than traditional columns used with electrospray.
Manufacturer:  Thermo Scientific™ 72105107582
 View more versions of this product
Catalog No. 03-051-656
Add to cart
Description
Description
Thermo Scientific™ BioBasic 18 Nanobore HPLC Columns offer greater signal to noise ratio than traditional columns used with electrospray at flowrates of nL/min. versus mL/min.
  • High sensitivity flexible silica capillary columns
  • Excellent for LC/MS applications
Features of BioBasic 18 Solid Phase
  • 300Å pore size for maximum performance with biomolecules
  • High peak capacity stationary phase
  • Outstanding reproducibility, efficiency and column lifetime
Ordering Information:
Unless otherwise specified, IntegraFrit and PicoFrit are sold in single-column units.
Specifications
Specifications
Reversed Phase
Silica
Reversed Phase
Nanobore Column with PicoFrit
Nanospray LC/MS
5 μm
300 Å
C18
Yes
9%
Picofrit Multi
0.075 mm
100 mm
2 to 10
L1
BioBasic