Thermo Scientific™ BioBasic™ 4 LC Columns Available on GSA/VA Contract for Federal Government customers only.

BioBasic 4 LC Columns are designed for the analysis of larger proteins and peptides.

Manufacturer: Thermo Scientific™ 72305-011001

 View all options for this product

Catalog No. 72305011001

  Check Availability
Add to Cart


    Column Type Reversed Phase
    Type Guard Cartridge, 4 PK
    Carbon Load 4%
    USP Type L26
    Endcapped Yes
    Max. Pressure 400bar
    Particle Size 5µm
    Temperature 60°C
    Pore Size 300Å
    pH 2 to 9
    Stationary Phase C4
    Surface Area 100m²/g
    Length (Metric) 10mm
    Column Format Guard Cartridge, 4 PK
    Diameter (Metric) 1mm

    Designed for the analysis of larger proteins and peptides, Thermo Scientific™ BioBasic™ 4 LC Columns are high-performance columns ideal for the analysis of proteins, peptides and other biomolecules with high hydrophobicity.

    • Based on 300Å silica for outstanding biomolecule performance
    • Lower carbon load for optimal retention of larger peptides and proteins
    • Outstanding reproducibility, efficiency and column lifetime
    • Excellent for LC/MS separations

    Ordering Information:

    Other column dimensions are available, including preparative columns. Please call your Customer Service Representative for more information.

  • Description & Specifications
  • Accessories

Related Products

Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit